Order Entry
Switzerland
ContactUsLinkComponent
Anti-Glucose Transporter-3 Rabbit Polyclonal Antibody
Anti-Glucose Transporter-3 Rabbit Polyclonal Antibody
Catalog # 400062-50
Anti-Glucose Transporter-3 Rabbit Polyclonal Antibody
Catalog # 400062-50

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glucose Transporter 3
  • Antigen symbol:
    GLUT3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: Glucose Transporter-3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human