Anti-G i alpha-1-Subunit, Internal (159-168) Rabbit Polyclonal Antibody
Codice catalogo 371720-50
Nome del fornitore: Merck Millipore (Oncogene)
Specifiche
- Antibody type:Primary
- Antigen name:G i alpha-1-Subunit
- Antigen symbol:GIA-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Epitope:internal
- Amino acid number:159 to 168
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: G i alpha-1-Subunit, Internal (159-168)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse