Anti-Complement 5b-9 Rabbit Polyclonal Antibody
Catalog # 204903-1
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:Complement 5b-9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Complement 5b-9
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: