Anti-NF68 Mouse Monoclonal Antibody [clone: NF-68]
Catalog # BSBTMA1070
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:NF68
- Clonality:Monoclonal
- Clone:NF-68
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Pig,Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Pig spinal cord.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: NF68
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig, Rat