Anti-AIM2 Mouse Monoclonal Antibody [clone: 10M5G5]
Catalog # NOVUNBP2-27354
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:absent in melanoma 2
- Antigen symbol:AIM2
- Clonality:Monoclonal
- Clone:10M5G5
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Gene ID:9447
- Antigen synonyms:PYHIN4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: AIM2 (absent in melanoma 2)
Clonality: monoclonal
Clone: 10M5G5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human