Anti-SEMA4D Mouse Monoclonal Antibody [clone: A8]
Catalog # NOVUNBP2-25198
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Semaphorin 4D
- Antigen symbol:SEMA4D
- Clonality:Monoclonal
- Clone:A8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Gene ID:10507
- Antigen synonyms:C9orf164|M-sema-G|SEMAJ|CD100|coll-4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: SEMA4D (sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D)
Clonality: monoclonal
Clone: A8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human