Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-UBA6 Rabbit Polyclonal Antibody
  NOVUNBP2-24587
undefined
Anti-UBA6 Rabbit Polyclonal Antibody
  NOVUNBP2-24587

 

  • Antibody type:
    Primary
  • Antigen name:
    ubiquitin-like modifier activating enzyme 6
  • Antigen symbol:
    UBA6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Gene ID:
    55236
  • Antigen synonyms:
    E1-L2|MOP-4|UBE1L2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: UBA6 (ubiquitin-like modifier activating enzyme 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: