Anti-SNX2 Rabbit Polyclonal Antibody
Katalognr. NOVUNBP2-24513
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:sorting nexin 2
- Antigen symbol:SNX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:6643
- Antigen synonyms:TRG-9
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: SNX2 (sorting nexin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: