Order Entry
Switzerland
ContactUsLinkComponent
Anti-MAP3K14 Rabbit Polyclonal Antibody (DyLight® 350)
Anti-MAP3K14 Rabbit Polyclonal Antibody (DyLight® 350)
N° de catalogue NOVUNBP2-23603UV
Fournisseur:  Novus Biologicals
Anti-MAP3K14 Rabbit Polyclonal Antibody (DyLight® 350)
N° de catalogue NOVUNBP2-23603UV
Fournisseur:  Novus Biologicals

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase kinase kinase 14
  • Antigen symbol:
    MAP3K14
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 350
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    9020
  • Antigen synonyms:
    FTDCR1B|HS|NIK|HSNIK
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Spécifications

À propos de cet article

Type: Primary
Antigen: MAP3K14 (mitogen-activated protein kinase kinase kinase 14)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 350
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse