Anti-MAP3K14 Rabbit Polyclonal Antibody (DyLight® 650)
Katalognr. NOVUNBP2-23603C
Anbieter: Novus Biologicals
Technische Daten
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase kinase 14
- Antigen symbol:MAP3K14
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:9020
- Antigen synonyms:FTDCR1B|HS|NIK|HSNIK
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: MAP3K14 (mitogen-activated protein kinase kinase kinase 14)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse