Anti-ZEB1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2A8A6]
Catalog # NOVUNBP2-23484PE
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:zinc finger E-box binding homeobox 1
- Antigen symbol:ZEB1
- Clonality:Monoclonal
- Clone:2A8A6
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:6935
- Antigen synonyms:ZFHEP|TCF8|FECD6|PPCD3|ZFHX1A|NIL2A|DELTAEF1|BZP|AREB6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: ZEB1 (zinc finger E-box binding homeobox 1)
Clonality: monoclonal
Clone: 2A8A6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human