Anti-IL7 Receptor alpha Mouse Monoclonal Antibody [clone: 22F3D9]
Catalog # NOVUNBP2-22376
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 7 Receptor
- Antigen symbol:IL7R
- Clonality:Monoclonal
- Clone:22F3D9
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b lambda
- Reactivity:Human
- Western blot:Yes
- Gene ID:3575
- Antigen synonyms:IL7RA|CD127|ILRA|CDW127|IL-7R-alpha
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: IL7 Receptor alpha
Clonality: monoclonal
Clone: 22F3D9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Lambda
Reactivity: Human