Anti-RTEL1 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP2-22360R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Regulator of telomere elongation helicase 1
- Antigen symbol:RTEL1
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:51750
- Antigen synonyms:DKCA4|NHL|RTEL|DKCB5|PFBMFT3|C20orf41
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: RTEL1 (regulator of telomere elongation helicase 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human