Order Entry
Switzerland
ContactUsLinkComponent
Anti-APP Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: MOAB-2]
Anti-APP Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: MOAB-2]
Catalog # NOVUNBP2-13075H
Supplier:  Novus Biologicals
undefined
Anti-APP Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: MOAB-2]
Catalog # NOVUNBP2-13075H
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Amyloid beta (A4) Precursor Protein
  • Antigen symbol:
    APP
  • Clonality:
    Monoclonal
  • Clone:
    MOAB-2
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    351
  • Antigen synonyms:
    CTFgamma|CVAP|ABETA|APPI|AD1|PN-II|ABPP|AAA|PN2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone: MOAB-2
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat