Anti-EBI3 Rat Monoclonal Antibody [clone: 10J706]
Catalog # NOVUNBP2-03938
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Epstein-Barr virus induced 3
- Antigen symbol:EBI3
- Clonality:Monoclonal
- Clone:10J706
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Western blot:Yes
- Gene ID:10148
- Antigen synonyms:IL35 subunit|IL-27B|EBV-induced gene 3 protein|IL27 subunit|Epstein-Barr virus induced gene 3|interleukin-27 subunit beta|cytokine receptor|IL-27 subunit beta|epstein-Barr virus-induced gene 3 protein|IL27B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: EBI3 (Epstein-Barr virus induced 3)
Clonality: monoclonal
Clone: 10J706
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse