Anti-SUV420H1 Rabbit Polyclonal Antibody (DyLight® 650)
NOVUNBP1-97312C
- Antibody type:Primary
- Antigen name:Suppressor of Variegation 4-20 Homolog 1
- Antigen symbol:SUV420H1
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Gene ID:51111
- Antigen synonyms:CGI85|CGI-85|KMT5B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: SUV420H1 (suppressor of variegation 4-20 homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse