Anti-GLP1R Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-97308R
Anbieter: Novus Biologicals
Technische Daten
- Antibody type:Primary
- Antigen name:Glucagon-like peptide 1 receptor
- Antigen symbol:GLP1R
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:2740
- Antigen synonyms:GLP-1-R|GLP-1|Glucagon-like peptide-1 receptor|GLP-1 receptor|GLP-1R
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: GLP1R (glucagon-like peptide 1 receptor)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse