Anti-UBE2E1 Rabbit Polyclonal Antibody (Biotin)
NOVUNBP1-78983B
- Antibody type:Primary
- Antigen name:Ubiquitin-conjugating enzyme E2E 1
- Antigen symbol:UBE2E1
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Gene ID:7324
- Antigen synonyms:UB2E1_HUMAN|UbcH6|Ubiquitin-conjugating enzyme E2 E1|Ubiquitin carrier protein E1|Ubiquitin-protein ligase E1.|UBE2E1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: UBE2E1 (ubiquitin-conjugating enzyme E2E 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse