Anti-TLR4 Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNBP1-78427C
Anbieter: Novus Biologicals
Technische Daten
- Antibody type:Primary
- Antigen name:Toll-like Receptor 4
- Antigen symbol:TLR4
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:7099
- Antigen synonyms:TLR4|Toll-like receptor 4|Rasl2-8|TLR-4|Lps|Ran/M1|Ly87|hToll|CD284|ARMD10|TOLL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: TLR4 (toll-like receptor 4)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat