Anti-TLR4 Rabbit Polyclonal Antibody
The TLR4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TLR4. This antibody reacts with human, mouse, rat. The TLR4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:50
Type: Primary
Antigen: TLR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- NOVUNBP1-78427SS
- NOVUNBP1-78427
- Antigen symbol:
- TLR4
- TLR4
- Antigen name:
- Toll-like Receptor 4
- Toll-like Receptor 4
- Antigen synonyms:
- TLR4|Toll-like receptor 4|Rasl2-8|TLR-4|Lps|Ran/M1|Ly87|hToll|CD284|ARMD10|TOLL
- TLR4|Toll-like receptor 4|Rasl2-8|TLR-4|Lps|Ran/M1|Ly87|hToll|CD284|ARMD10|TOLL
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 7099
- 7099
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together