Anti-GSAP Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNBP1-78400C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Gamma-Secretase Activating Protein
- Antigen symbol:GSAP
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:54103
- Antigen synonyms:PION|Protein pigeon homolog|GSAP|Gamma-secretase-activating protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: GSAP (gamma-secretase activating protein)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human