Anti-SERF1A Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNBP1-78393V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Small EDRK-rich Factor 1A
- Antigen symbol:SERF1A
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Gene ID:8293
- Antigen synonyms:H4F5|SERF1|FAM2A|4F5|SMAM1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SERF1A (small EDRK-rich factor 1A (telomeric))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human