Anti-ABCB5 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNBP1-77687V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:ATP-binding cassette, sub-family B (MDR/TAP), member 5
- Antigen symbol:ABCB5
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Gene ID:340273
- Antigen synonyms:ABCB5alpha|EST422562|ABCB5beta
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: ABCB5 (ATP-binding cassette, sub-family B (MDR/TAP), member 5)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse