Anti-TPH2 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: TPH2 (tryptophan hydroxylase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Specifications
- Catalog No:
- NOVUNBP1-77559SS
- NOVUNBP1-77559
- Antigen-Symbol:
- TPH2
- TPH2
- Antigen-Name:
- Tryptophan hydroxylase 2
- Tryptophan hydroxylase 2
- Antigen-Synonyme:
- tryptophan 5-hydroxylase 2|neuronal tryptophan hydroxylase|tryptophan 5-monooxygenase 2|NTPH|ADHD7
- tryptophan 5-hydroxylase 2|neuronal tryptophan hydroxylase|tryptophan 5-monooxygenase 2|NTPH|ADHD7
- Antikörper-Typ:
- Primär
- Primär
- Klonalität:
- Polyklonal
- Polyklonal
- Konjugation:
- Unconjugated
- Unconjugated
- Reaktivität:
- Human
- Human
- Wirt:
- Rabbit
- Rabbit
- Gen-ID:
- 121278
- 121278
- Western blot:
- Yes
- Yes
- Lagertemperatur:
- Cool
- Cool
- Transporttemperatur:
- Ice
- Ice
Specifications
Frequently Bought Together