Anti-NBR1 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNBP1-71703G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Neighbor of BRCA1 gene 1
- Antigen symbol:NBR1
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Gene ID:4077
- Antigen synonyms:mKIAA0049
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: NBR1 (neighbor of BRCA1 gene 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse