Anti-PTCH1 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNBP1-71662V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:patched 1
- Antigen symbol:PTCH1
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:5727
- Antigen synonyms:PTCH|HPE7|PTCH11|PTC|PTC1|NBCCS|BCNS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: PTCH1 (patched 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human