Order Entry
Switzerland
ContactUsLinkComponent
Anti-OPA1 Mouse Monoclonal Antibody [clone: 1E8-1D9]
Anti-OPA1 Mouse Monoclonal Antibody [clone: 1E8-1D9]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-OPA1 Mouse Monoclonal Antibody [clone: 1E8-1D9]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-71656SS
  • NOVUNBP1-71656
  • Antigen symbol:
  • OPA1
  • OPA1
  • Antigen name:
  • Optic Atrophy 1
  • Optic Atrophy 1
  • Antigen synonyms:
  • optic atrophy protein 1|mitochondrial dynamin-like 120 kDa protein|largeG|NTG|mitochondrial|MGM1|dynamin-like guanosine triphosphatase|dynamin-like 120 kDa protein|NPG|mitochondrial dynamin-like GTPase
  • optic atrophy protein 1|mitochondrial dynamin-like 120 kDa protein|largeG|NTG|mitochondrial|MGM1|dynamin-like guanosine triphosphatase|dynamin-like 120 kDa protein|NPG|mitochondrial dynamin-like GTPase
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 1E8-1D9
  • 1E8-1D9
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 4976
  • 4976
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together