Anti-mtTFA Mouse Monoclonal Antibody (DyLight® 405) [clone: 18G102B2E11]
Katalognr. NOVUNBP1-71648V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Transcription Factor A, Mitochondrial
- Antigen symbol:TFAM
- Clonality:Monoclonal
- Clone:18G102B2E11
- Conjugation:DyLight® 405
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human,Rat
- Western blot:Yes
- Gene ID:7019
- Antigen synonyms:TCF6L1|TCF6L2|MTTFA|TCF6L3|TCF6|MTTF1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: mtTFA
Clonality: monoclonal
Clone: 18G102B2E11
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Rat