Anti-TEK Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNBP1-69753B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Angiopoietin-1 Receptor
- Antigen symbol:TEK
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:7010
- Antigen synonyms:Angiopoietin-1 receptor,EC=2.7.10.1 Tunica interna endothelial cell kinase Tyrosine-protein kinase receptor TEK Tyrosine-protein kinase receptor TIE-2 hTIE2 p140 TEK CD202b TEK TIE2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: TEK (TEK tyrosine kinase, endothelial)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse