Anti-STK33 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-54156R
Supplier: Novus Biologicals
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Serine/threonine Kinase 33
- Antigen-Symbol:STK33
- Klonalität:Polyklonal
- Konjugation:DyLight® 550
- Wirt:Rabbit
- ImmunFluoreszenz:Yes
- Reaktivität:Human
- Gen-ID:65975
- Antigen-Synonyme:EC 2.7.11
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:100 µL
- VE:0,1 mL
Specifications
About this item
The STK33 Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to STK33. This antibody reacts with human, mouse. The STK33 Antibody [DyLight 550] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: STK33
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse