Anti-STK33 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NOVUNBP1-54156H
Supplier: Novus Biologicals
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Serine/threonine Kinase 33
- Antigen-Symbol:STK33
- Klonalität:Polyklonal
- Konjugation:HRP (Horseradish Peroxidase)
- Wirt:Rabbit
- ImmunFluoreszenz:Yes
- Reaktivität:Human
- Gen-ID:65975
- Antigen-Synonyme:EC 2.7.11
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:100 µL
- VE:0,1 mL
Specifications
About this item
Type: Primary
Antigen: STK33 (serine/threonine kinase 33)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse