Order Entry
Switzerland
ContactUsLinkComponent
Anti-NKX6-1 Rabbit Polyclonal Antibody (Biotin)
Anti-NKX6-1 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNBP1-49672B
Supplier:  Novus Biologicals
undefined
Anti-NKX6-1 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNBP1-49672B
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    NK6 homeobox 1
  • Antigen symbol:
    NKX6-1
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Gene ID:
    4825
  • Antigen synonyms:
    NKX6.1|NKX6A
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

The NKX6.1 Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to NKX6.1. This antibody reacts with human, mouse. The NKX6.1 Antibody [Biotin] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:100

Type: Primary
Antigen: NKX6.1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse