Anti-LDHA Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNBP1-48336C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Lactate Dehydrogenase A
- Antigen symbol:LDHA
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:3939
- Antigen synonyms:LDHM|PIG19|LDH1|HEL-S-133P|GSD11
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: LDHA (lactate dehydrogenase A)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse