Anti-ESRRA Rabbit Polyclonal Antibody (DyLight® 550)
NOVUNBP1-47254R
- Antibody type:Primary
- Antigen name:Estrogen Related Receptor alpha
- Antigen symbol:ERR-alpha
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:2101
- Antigen synonyms:NR3B1|ESRL1 |ERRa |ERRalpha|ERR1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: ESRRA (estrogen-related receptor alpha)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse