Anti-SNX27 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNBP1-45283B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Sorting Nexin 27
- Antigen symbol:SNX27
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:81609
- Antigen synonyms:MRT1|MY014
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SNX27 (sorting nexin family member 27)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse