Anti-SULF2 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 2B4]
NOVUNBP1-36727F
- Antibody type:Primary
- Antigen name:sulfatase 2
- Clonality:Monoclonal
- Clone:2B4
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:55959
- Antigen synonyms:HSULF-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: SULF2 (sulfatase 2)
Clonality: monoclonal
Clone: 2B4
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse