Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-TMEM97 Rabbit Polyclonal Antibody
 
undefined
Anti-TMEM97 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNBP1-30437
  • NOVUNBP1-30437SS
  • Antigen symbol:
  • TMEM97
  • TMEM97
  • Antigen name:
  • Transmembrane Protein 97
  • Transmembrane Protein 97
  • Antigen synonyms:
  • MAC30
  • MAC30
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 27346
  • 27346
  • Western blot:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together