Anti-Palladin Mouse Monoclonal Antibody (DyLight® 488) [clone: 1E6]
Catalog # NOVUNBP1-25959G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Palladin
- Clonality:Monoclonal
- Clone:1E6
- Conjugation:DyLight® 488
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:23022
- Antigen synonyms:SIH002|CGI-151|MYN|CGI151|PNCA1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Palladin
Clonality: monoclonal
Clone: 1000000
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Mouse, Rat