Anti-SPHK1 Rabbit Polyclonal Antibody (Biotin)
NOVUNBP1-22974B
- Antibody type:Primary
- Antigen name:Sphingosine Kinase 1
- Antigen symbol:SPK1
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:8877
- Antigen synonyms:SK 1|sphk|SPK1|SPK-1|SK1|SPK 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: SPHK1 (sphingosine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human