Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 488) [clone: OC2-1D11]
Catalog # NOVUNBP1-18963G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Oval Cell Marker
- Clonality:Monoclonal
- Clone:OC2-1D11
- Conjugation:DyLight® 488
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-1D11
Conjugation: DyLight® 488
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse