Order Entry
Switzerland
ContactUsLinkComponent
Anti-HPd1 Mouse Monoclonal Antibody [clone: DHIC2-4A10]
Anti-HPd1 Mouse Monoclonal Antibody [clone: DHIC2-4A10]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-HPd1 Mouse Monoclonal Antibody [clone: DHIC2-4A10]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-18953SS
  • NOVUNBP1-18953
  • Antigen symbol:
  • HPD1
  • HPD1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • DHIC2-4A10
  • DHIC2-4A10
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together