Anti-PDK4 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNBP1-07049F
Anbieter: Novus Biologicals
Technische Daten
- Antibody type:Primary
- Antigen name:pyruvate dehydrogenase kinase, isozyme 4
- Antigen symbol:PDK4
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:5166
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: PDK4 (pyruvate dehydrogenase kinase, isozyme 4)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human