Anti-PDK4 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-07047R
Anbieter: Novus Biologicals
Technische Daten
- Antibody type:Primary
- Antigen name:Pyruvate Dehydrogenase Kinase 4
- Antigen symbol:PDK4
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:5166
- Antigen synonyms:FLJ40832|Pyruvate dehydrogenase kinase isoform 4|Pyruvate dehydrogenase kinase isozyme 4 mitochondrial.|Pyruvate dehydrogenase [lipoamide] kinase isozyme 4 mitochondrial|Pyruvate dehydrogenase kinase 4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: PDK4 (pyruvate dehydrogenase kinase, isozyme 4)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse