Anti-DIO3 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNBP1-05767G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:deiodinase, iodothyronine, type III
- Antigen symbol:DIO3
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Gene ID:1735
- Antigen synonyms:TXDI3|DIOIII|D3|5DIII
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: DIO3 (deiodinase, iodothyronine, type III)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: