Anti-Ethenoadenosine Mouse Monoclonal Antibody (DyLight® 488) [clone: 1G4]
Catalog # NOVUNB600-442G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Ethenoadenosine
- Clonality:Monoclonal
- Clone:1G4
- Conjugation:DyLight® 488
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2 lambda
- Reactivity:Multiple
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Ethenoadenosine
Clonality: monoclonal
Clone: 1G4
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG2 Lambda
Reactivity: Pan