Anti-BIRC5 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 60.11]
Catalog # NOVUNB500-238F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:baculoviral IAP repeat containing 5
- Antigen symbol:BIRC5
- Clonality:Monoclonal
- Clone:60.11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:332
- Antigen synonyms:API4|EPR-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: monoclonal
Clone: 60.11
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat