Anti-POT1 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB500-176V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Protection of telomeres 1
- Antigen symbol:POT1
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:25913
- Antigen synonyms:HPOT1|CMM10
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: POT1 (protection of telomeres 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human