Anti-ABCE1 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNB400-116B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, Subfamily E, Member 1
- Antigen symbol:ABCE1
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:6059
- Antigen synonyms:RNase 4 inhibitor|Ribonuclease 4 Inhibitor|ABC 38|ABCE 1|OABP|ATP Binding Cassette Sub Family E Member 1|RLI|RNS4I|ABC38|RNase L Inhibitor
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: ABCE1 (ATP-binding cassette, sub-family E (OABP), member 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human