Anti-NES Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10C2]
Catalog # NOVUNB300-266PE
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Nestin
- Antigen symbol:NES
- Clonality:Monoclonal
- Clone:10C2
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Gene ID:10763
- Antigen synonyms:Nbla00170
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: NES (nestin)
Clonality: monoclonal
Clone: 10C2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human