Anti-NES Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 10C2]
Catalog # NOVUNB300-266H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Nestin
- Antigen symbol:NES
- Clonality:Monoclonal
- Clone:10C2
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:10763
- Antigen synonyms:FLJ21841|C78523|ESTM46|AA166324|Nbla00170
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: NES (nestin)
Clonality: monoclonal
Clone: 10C2
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human